missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLCO5A1 Antibody (S02-5J4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19774-100ul
This item is not returnable.
View return policy
Description
SLCO5A1 Monoclonal antibody specifically detects SLCO5A1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SLCO5A1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 81796 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S02-5J4 | |
| Unconjugated | |
| FLJ39560, member 15, OATP5A1, OATPRP4, organic anion transporter polypeptide-related protein 4, SLC21A15, solute carrier organic anion transporter family, member 5A1 | |
| Recombinant protein of human SLCO5A1 (Uniprot # Q9H2Y9) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction