missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLCO4C1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06278PCP
This item is not returnable.
View return policy
Description
SLCO4C1 Polyclonal antibody specifically detects SLCO4C1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SLCO4C1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| PRO2176, solute carrier family 21 member 20, solute carrier organic anion transporter family, member 4C1 | |
| Produced in rabbits immunized with E. coli-derived Human SLCO4C1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 353189 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction