missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLCO4A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556536
This item is not returnable.
View return policy
Description
Immunogen sequence: GYPRQLPGSQ RYAVMRAAEM HQLKDSSRGE ASNPDFGKTI RDLPL Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 78%.
Mediates the Na(+)-independent transport of organic anions such as the thyroid hormones T3 (triiodo-L-thyronine), T4 (thyroxine) and rT3, and of estrone-3-sulfate and taurocholate.
Specifications
| SLCO4A1 | |
| Polyclonal | |
| Unconjugated | |
| SLCO4A1 | |
| colon organic anion transporter; OATP1; OATP4A1; OATPE; OATP-E; OATPRP1; OATP-RP1; Organic anion transporter polypeptide-related protein 1; organic anion transporting polypeptide E; organic anion-transporting polypeptide E; POAT; SLC21A12; SLCO4A1; Sodium-independent organic anion transporter E; solute carrier family 21 (organic anion transporter), member 12; solute carrier family 21 member 12; Solute carrier organic anion transporter family member 4A1; solute carrier organic anion transporter family, member 4A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28231 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96BD0 | |
| SLCO4A1 | |
| Recombinant protein corresponding to Human SLCO4A1. Recombinant protein control fragment (Product #RP-95579). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction