missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLCO3A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59678
This item is not returnable.
View return policy
Description
SLCO3A1 Polyclonal specifically detects SLCO3A1 in Human samples. It is validated for Western Blot.
Specifications
| SLCO3A1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ40478, OATP3A1OATP-RP3, OATP-DOATPRP3, OATPDSLC21A11, Organic anion transporter polypeptide-related protein 3, Organic anion-transporting polypeptide D, PGE1 transporter, Sodium-independent organic anion transporter D, solute carrier family 21 (organic anion transporter), member 11, Solute carrier family 21 member 11, solute carrier organic anion transporter family member 3A1, solute carrier organic anion transporter family, member 3A1 | |
| Rabbit | |
| 76 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9UIG8 | |
| SLCO3A1 | |
| Synthetic peptides corresponding to SLCO3A1(solute carrier organic anion transporter family, member 3A1) The peptide sequence was selected from the middle region of SLCO3A1 (NP_037404). Peptide sequence MEIAVVAGFAAFLGKYLEQQFNLTTSSANQLLGMTAIPCACLGIFLGGLL The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 28232 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction