missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLCO1C1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93140-0.1ml
This item is not returnable.
View return policy
Description
SLCO1C1 Polyclonal antibody specifically detects SLCO1C1 in Human samples. It is validated for Western Blot
Specifications
| SLCO1C1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| OATP1, OATP14Thyroxine transporter, OATP1C1Solute carrier family 21 member 14, OATPFOAT-RP-5, OATP-FOrganic anion-transporting polypeptide 14, OATPRP5, Organic anion transporter F, Organic anion transporter polypeptide-related protein 5, organic anion transporting polypeptide 14, SLC21A14OATP-14, solute carrier family 21 (organic anion transporter), member 14, solute carrier organic anion transporter family member 1C1, solute carrier organic anion transporter family, member 1C1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-554 of human SLCO1C1 (Q9NYB5). SDVAGLTVSYQGTKPVSYHERALFSDCNSRCKCSETKWEPMCGENGITYVSACLAGCQTSNRSGKNIIFYNCTCVGIAASKSGNSSGIVGRCQKDNGCPQMFLYF | |
| 0.1 mL | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 53919 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction