missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLCO1A2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94661-0.02ml
This item is not returnable.
View return policy
Description
SLCO1A2 Polyclonal antibody specifically detects SLCO1A2 in Rat samples. It is validated for Western Blot
Specifications
| SLCO1A2 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000 | |
| OATP1, OATP1A2member 3, OATP-AOATP-1, OATPSolute carrier family 21 member 3, Organic anion-transporting polypeptide 1, Sodium-independent organic anion transporter, solute carrier organic anion transporter family member 1A2, solute carrier organic anion transporter family, member 1A2 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human SLCO1A2 (NP_066580.1). SYEGIPQDLYVENDIFADC | |
| 0.02 mL | |
| Cancer, Cell Biology, Endocrinology, Immunology, Signal Transduction | |
| 6579 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction