missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC9A9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13718-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a sodium hydrogen exchanger that is a member of the solute carrier 9 protein family. The encoded protein localizes the to the late recycling endosomes and may play an important role in maintaining cation homeostasis. Defects in this gene are associated with attention-deficit/hyperactivity disorder.Specifications
| SLC9A9 | |
| Polyclonal | |
| Unconjugated | |
| SLC9A9 | |
| Na(+)/H(+) exchanger 9, Nbla00118, NHE 9, NHE9, SLC9A9, Sodium/hydrogen exchanger 9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 285195, 331004, 363115 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BZ00, Q8IVB4 | |
| Slc9a9 | |
| SLC9A9 Fusion Protein Ag4660 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction