missing translation for 'onlineSavingsMsg'
Learn More

SLC9A9 Mouse anti-Human, Mouse, Rat, Clone: 1B8D9, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen SLC9A9
Clone 1B8D9
Concentration 1 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16808585
View Documents
Proteintech
66577-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16898575
View Documents
Proteintech
66577-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a sodium hydrogen exchanger that is a member of the solute carrier 9 protein family. The encoded protein localizes the to the late recycling endosomes and may play an important role in maintaining cation homeostasis. Defects in this gene are associated with attention-deficit/hyperactivity disorder.
Specifications

Specifications

SLC9A9
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Na(+)/H(+) exchanger 9, Nbla00118, NHE 9, NHE9, SLC9A9, Sodium/hydrogen exchanger 9
Slc9a9
IgG2a
Protein A
Antibody
1B8D9
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
Unconjugated
Mouse
Mouse, Human, Rat
Q8BZ00, Q8IVB4
285195, 331004, 363115
SLC9A9 Fusion Protein Ag4660
Primary
-20°C
SLC9A9
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.