missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC9A8 Monoclonal Antibody (7A11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA146357
This item is not returnable.
View return policy
Description
Do not boil samples prior to Western Blot. Suggested positive control: antigen standard for SLC9A8 (transient overexpression lysate), HeLa whole cell lysate..
Sodium-hydrogen exchangers (NHEs), such as SLC9A8, are integral transmembrane proteins that exchange extracellular Na+ for intracellular H+. NHEs have multiple functions, including intracellular pH homeostasis, cell volume regulation, and electroneutral NaCl absorption in epithelia (Xu et al., 2008).
Specifications
| SLC9A8 | |
| Monoclonal | |
| 1.2 mg/mL | |
| TBS with 0.05% sodium azide | |
| Q4L208, Q8R4D1, Q9Y2E8 | |
| SLC9A8 | |
| A fusion protein containing the C-terminal 89 residues of human NHE8. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 7A11 | |
| Unconjugated | |
| SLC9A8 | |
| 1200006P13Rik; 6430709P13Rik; AI182282; DKFZp686C03237; fj61b02; FLJ42500; hypothetical protein LOC494043; KIAA0939; MGC138418; Na(+)/H(+) exchanger 8; Nhe8; NHE-8; NHE-8 antibody; SLC9A8; SLC9A8 antibody; sodium hydrogen exchanger 8; sodium/hydrogen exchanger 8; solute carrier family 9 (sodium/hydrogen exchanger), isoform 8; solute carrier family 9 (sodium/hydrogen exchanger), member 8; solute carrier family 9 member 8; solute carrier family 9 member A8; solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8; wu:fj61b02; zgc:103660 | |
| Mouse | |
| Protein G | |
| RUO | |
| 23315, 311651, 77031 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction