missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC9A7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62455
This item is not returnable.
View return policy
Description
SLC9A7 Polyclonal specifically detects SLC9A7 in Human samples. It is validated for Western Blot.
Specifications
| SLC9A7 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| SLC9A7 | |
| Synthetic peptides corresponding to SLC9A7(solute carrier family 9 (sodium/hydrogen exchanger), member 7) The peptide sequence was selected from the N terminal of SLC9A7. Peptide sequence LGWGLRVAAAASASSSGAAAEDSSAMEELATEKEAEESHRQDSVSLLTFI. | |
| Affinity purified | |
| RUO | |
| 84679 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Na(+)/H(+) exchanger 7, NHE-7, NHE7SLC9A6, nonselective sodium potassium/proton exchanger, sodium/hydrogen exchanger 7, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 7, solute carrier family 9 (sodium/hydrogen exchanger), member 7, Solute carrier family 9 member 7 | |
| Rabbit | |
| 80 kDa | |
| 100 μL | |
| Primary | |
| Goat: 85%. | |
| Human, Mouse, Canine, Guinea Pig, Goat, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction