missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC9A6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550139
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SLC9A6 / NHE-6 protein.
Na+/H+ exchangers-1-6 (Na+/H+ antiporters, NHE-1-6) are integral membrane proteins that are expressed in most mammalian tissues where they regulate intracellular pH and cell volume. NHEs mediate the secondary active extrusion of hydrogen (H+) ions out of cells in exchange for extracellular sodium (Na+). Excluding NHE-1, which is ubiquitously expressed, the NHE isoforms 2-6 have distinct tissue- and cell type-dependent expression, and inhibitiory characteristics by amiloride analogs.
Specifications
| SLC9A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC9A6 | |
| 3732426M05; 6430520C02Rik; KIAA0267; mKIAA0267; MRSA; Na(+)/H(+) exchanger 6; NHE6; NHE-6; RGD1563582; SLC9A6; Sodium/hydrogen exchanger 6; solute carrier family 9 (sodium/hydrogen exchanger), isoform 6; solute carrier family 9 (sodium/hydrogen exchanger), member 6; Solute carrier family 9 member 6; solute carrier family 9 member A6; solute carrier family 9, subfamily A (NHE6, cation proton antiporter 6), member 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10479, 236794 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92581 | |
| SLC9A6 | |
| Synthesized peptide derived from internal of human SLC9A6. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction