missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC9A5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577561
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Na+/H+ exchangers-1-6 (Na+/H+ antiporters, NHE-1-6) are integral membrane proteins that are expressed in most mammalian tissues, where they regulate intracellular pH and cell volume. NHEs mediate the secondary active extrusion of hydrogen (H+) ions out of cells in exchange for extracellular sodium (Na+). Excluding NHE-1, which is ubiquitously expressed, NHE isoforms 2-6 have distinct tissue- and cell type-dependent expression and inhibitiory characteristics by amiloride analogs. Human NHE-5 transcript (solute carrier family 9, isoform-5, SLC9A5) is an 896 amino acid protein that is expressed in brain, testis, spleen and skeletal muscle.
Specifications
| SLC9A5 | |
| Polyclonal | |
| Unconjugated | |
| SLC9A5 | |
| Na+/H+ Exchanger 5 (NHE-5) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 192215, 277973, 6553 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q14940, Q9Z0X2 | |
| Slc9a5 | |
| Peptide CAEKELPWKSGQGD, corresponding to amino acid residues 759- 772 of rat NHE-5 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction