missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC9A4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£143.00 - £396.00
Specifications
| Antigen | SLC9A4 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18213704
|
Novus Biologicals
NBP1-46583-0.025ML |
0.025 mL |
£143.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18259194
|
Novus Biologicals
NBP1-46583 |
0.1 mL |
£396.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC9A4 Polyclonal specifically detects SLC9A4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| SLC9A4 | |
| Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| 389015 | |
| IgG | |
| Unpurified | |
| Specific for SLC9A4. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), member 4 | |
| SLC9A4 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title