Learn More
SLC9A3R1 Mouse anti-Human, Clone: EBP-10, Abnova™
Mouse monoclonal antibody raised against human SLC9A3R1.
Brand: Abnova MAB12530.50ug
Description
This gene encodes a sodium/hydrogen exchanger regulatory cofactor. The protein interacts with and regulates various proteins including the cystic fibrosis transmembrane conductance regulator and G-protein coupled receptors such as the beta2-adrenergic receptor and the parathyroid hormone 1 receptor. The protein also interacts with proteins that function as linkers between integral membrane and cytoskeletal proteins. The protein localizes to actin-rich structures including membrane ruffles, microvilli, and filopodia. Mutations in this gene result in hypophosphatemic nephrolithiasis/osteoporosis type 2, and loss of heterozygosity of this gene is implicated in breast cancer
Specifications
| SLC9A3R1 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation (2 ug/mL) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| EBP50/NHERF/NHERF1/NPHLOP2 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 9368 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
| Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| EBP-10 | |
| Mouse monoclonal antibody raised against human SLC9A3R1. | |
| In PBS, pH 7.4 (0.09% sodium azide). | |
| SLC9A3R1 | |
| Recombinant protein corresponding to human SLC9A3R1. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.