missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC9A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557163
This item is not returnable.
View return policy
Description
Immunogen sequence: SLRESIRKDS SLNREHRAST STSRYLSLPK NTKLPEKLQK RRTISIADGN SSDSDADAGT TVLNLQPRAR RFLPEQFSKK S Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 85%.
Involved in pH regulation to eliminate acids generated by active metabolism or to counter adverse environmental conditions. Major proton extruding system driven by the inward sodium ion chemical gradient. Seems to play an important role in colonic sodium absorption.
Specifications
| SLC9A2 | |
| Polyclonal | |
| Unconjugated | |
| Slc9a2 | |
| 2210416H12Rik; 4932415O19Rik; AV333884; H7; Na(+)/H(+) exchanger 2; Na/H ion exchanger; Nhe2; NHE-2; Slc9a2; Sodium/hydrogen exchanger 2; Solute carrier family 9 (sodium/hydrogen exchanger 2), antiporter 2, Na+/H+ (Na+/H+ exchanger 2); solute carrier family 9 (sodium/hydrogen exchanger), member 2; solute carrier family 9 member 2; solute carrier family 9 member A2; solute carrier family 9, member 2; solute carrier family 9, subfamily A (NHE2, cation proton antiporter 2), member 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6549 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBY0 | |
| Slc9a2 | |
| Recombinant protein corresponding to Human SLC9A2. Recombinant protein control fragment (Product #RP-102651). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction