missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC9A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115918
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SLC9A2.
Involved in pH regulation to eliminate acids generated by active metabolism or to counter adverse environmental conditions. Major proton extruding system driven by the inward sodium ion chemical gradient. Seems to play an important role in colonic sodium absorption.
Specifications
| SLC9A2 | |
| Polyclonal | |
| Unconjugated | |
| Slc9a2 | |
| 2210416H12Rik; 4932415O19Rik; AV333884; H7; Na(+)/H(+) exchanger 2; Na/H ion exchanger; Nhe2; NHE-2; Slc9a2; Sodium/hydrogen exchanger 2; Solute carrier family 9 (sodium/hydrogen exchanger 2), antiporter 2, Na+/H+ (Na+/H+ exchanger 2); solute carrier family 9 (sodium/hydrogen exchanger), member 2; solute carrier family 9 member 2; solute carrier family 9 member A2; solute carrier family 9, member 2; solute carrier family 9, subfamily A (NHE2, cation proton antiporter 2), member 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 226999, 24783, 6549 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P48763, Q3ZAS0, Q9UBY0 | |
| Slc9a2 | |
| A synthesized peptide derived from human SLC9A2(Accession Q9UBY0), corresponding to amino acid residues L624-T674. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction