missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC9A11 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5140028

Product Code. 17894540

  • £394.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LSLMYSITKG YIKSQEDAKL LIKQIAVCES IYQKLCEILE TNKQDAVKEL VLMEHEGRDV VIALKTKQAI RNVIAKALKN LTFLCSRGII DKHEV Highest antigen sequence indentity to the following orthologs: Mouse - 33%, Rat - 31%.

SLC9A11 is involved in pH regulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC9A11
Polyclonal
Unconjugated
Slc9c2
Na(+)/H(+) exchanger 11; NHE-11; RGD1565820; SLC9A11; SLC9C2; Sodium/hydrogen exchanger 11; Solute carrier family 9 member 11; Solute carrier family 9 member C2; solute carrier family 9 member C2 (putative); solute carrier family 9, isoform 11; solute carrier family 9, member 11; solute carrier family 9, member C2 (putative)
Rabbit
Antigen affinity chromatography
RUO
284525
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5TAH2
Slc9c2
Recombinant protein corresponding to Human SLC9C2. Recombinant protein control fragment (Product #RP-109700).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.