missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC7A9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550887
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SLC7A9 protein.
Involved in the high-affinity. sodium-independent transport of cystine and neutral and dibasic amino acids (system b(0.+)-like activity). Thought to be responsible for the high-affinity reabsorption of cystine in the kidney tubule.
Specifications
| SLC7A9 | |
| Polyclonal | |
| Unconjugated | |
| SLC7A9 | |
| b(0,+)AT; b(0,+)AT1; B(0,+)-type amino acid transporter 1; b>o>, + amino acid transporter; b>o>, +AT; BAT1; CSNU3; FLJ94301; glycoprotein-associated amino acid transporter b0,+AT1; glycoprotein-associated amino acid transporter b0,+AT1; SLC7A9; solute carrier family 7 (amino acid transporter light chain, bo,+ system), member 9; solute carrier family 7 (cationic amino acid transporter, y+ system), member 9; solute carrier family 7 (glycoprotein-associated amino acid transporter light chain, bo,+ system), member 9; solute carrier family 7 member 9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11136 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P82251 | |
| SLC7A9 | |
| Synthetic peptide corresponding to residues near the N terminal of human solute carrier family 7 (amino acid transporter light chain, bo,+ system), member 9. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction