missing translation for 'onlineSavingsMsg'
Learn More

SLC7A7 Antibody, Novus Biologicals™

Product Code. 18390329 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390329 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390329 Supplier Novus Biologicals Supplier No. H00009056B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

SLC7A7 Polyclonal antibody specifically detects SLC7A7 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH03062.1
Gene Alias LAT3, LPI, Monocyte amino acid permease 2, MOP-2, solute carrier family 7 (cationic amino acid transporter, y+ system), member 7, Solute carrier family 7 member 7, Y+L amino acid transporter 1, Y+LAT1, y+LAT-1y(+)L-type amino acid transporter 1
Host Species Mouse
Immunogen SLC7A7 (AAH03062.1, 1 a.a. - 511 a.a.) full-length human protein. MVDSTEYEVASQPEVETSPLGDGASPGPEQVKLKKEISLLNGVCLIVGNMIGSGIFVSPKGVLIYSASFGLSLVIWAVGGLFSVFGALCYAELGTTIKKSGASYAYILEAFGGFLAFIRLWTSLLIIEPTSQAIIAITFANYMVQPLFPSCFAPYAASRLLAAACICLLTFINCAYVKWGTLVQDIFTYAKVLALIAVIVAGIVRLGQGASTHFENSFEGSSFAVGDIALALYSALFSYSGWDTLNYVTEEIKNPERNLPLSIGISMPIVTIIYILTNVAYYTVLDMRDILASDAVAVTFADQIFGIFNWIIPLSVALSCFGGLNASIVAASRLFFVGSREGHLPDAICMIHVERFTPVPSLLFNGIMALIYLCVEDIFQLINYYSFSYWFFVGLSIVGQLYLRWKEPDRPRPLKLSVFFPIVFCLCTIFLVAVPLYSDTINSLIGIAIALSGLPFYFLIIRVPEHKRPLYLRRIVGSATRYLQVLCMSVAAEMDLEDGGEMPKQRDPKSN
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9056
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.