missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC7A5 Recombinant Rabbit Monoclonal Antibody (0P6E8)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558039
This item is not returnable.
View return policy
Description
Immunogen sequence: FAAAKLLALA LIILLGFVQI GKGDVSNLDP NFSFEGTKLD VGNIVLALYS GLFAYGGWNY LNFVTEEMIN PYRNLPLAII ISLPIVTLVY VLTNLAYFTT L.
Solute carrier family 7 member 5 also known as large neutral amino acids transporter small subunit 1 is a protein that in humans is encoded by the SLC7A5 gene.It is sodium-independent, high-affinity transport of large neutral amino acids such as phenylalanine, tyrosine, leucine, arginine and tryptophan, when associated with SLC3A2/4F2hc.It involved in cellular amino acid uptake and the transport of L-DOPA across the blood-brain barrier, and that of thyroid hormones triiodothyronine (T3) and thyroxine (T4) across the cell membrane in tissues such as placenta.Acts as an amino acid exchanger and plays a role in neuronal cell proliferation (neurogenesis) in brain.
Specifications
| SLC7A5 | |
| Recombinant Monoclonal | |
| 0.7 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q01650 | |
| SLC7A5 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human LAT1/SLC7A5 (NP_003477.4). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 0P6E8 | |
| Unconjugated | |
| SLC7A5 | |
| 4F2; 4F2 LC; 4F2 light chain; 4F2HC; 4F2LC; 4T2HC; CD98; CD98 light chain; CD98HC; CD98LC; D0H16S474E; D16S469E; E16; hLAT1; integral membrane protein E16; large neutral amino acids transporter 1; large neutral amino acids transporter small subunit 1; LAT1; LAT-1; L-type amino acid transporter 1; MDU1; MP hLAT1; MPE16; NACAE; Protein TA1; SLC7A5; sodium-independent neutral amino acid transporter LAT1; solute carrier family 7 (amino acid transporter light chain, L system), member 5; solute carrier family 7 (cationic amino acid transporter, y+ system), member 5; solute carrier family 7 member 5; Ta1; tumor-associated protein 1; y+ system cationic amino acid transporter | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8140 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction