missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC7A5 Recombinant Rabbit Monoclonal Antibody (0P6E8)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558040

 View more versions of this product

Product Code. 30282426

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: FAAAKLLALA LIILLGFVQI GKGDVSNLDP NFSFEGTKLD VGNIVLALYS GLFAYGGWNY LNFVTEEMIN PYRNLPLAII ISLPIVTLVY VLTNLAYFTT L.

Solute carrier family 7 member 5 also known as large neutral amino acids transporter small subunit 1 is a protein that in humans is encoded by the SLC7A5 gene.It is sodium-independent, high-affinity transport of large neutral amino acids such as phenylalanine, tyrosine, leucine, arginine and tryptophan, when associated with SLC3A2/4F2hc.It involved in cellular amino acid uptake and the transport of L-DOPA across the blood-brain barrier, and that of thyroid hormones triiodothyronine (T3) and thyroxine (T4) across the cell membrane in tissues such as placenta.Acts as an amino acid exchanger and plays a role in neuronal cell proliferation (neurogenesis) in brain.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC7A5
Recombinant Monoclonal
0.7 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q01650
SLC7A5
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human LAT1/SLC7A5 (NP_003477.4).
100 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
0P6E8
Unconjugated
SLC7A5
4F2; 4F2 LC; 4F2 light chain; 4F2HC; 4F2LC; 4T2HC; CD98; CD98 light chain; CD98HC; CD98LC; D0H16S474E; D16S469E; E16; hLAT1; integral membrane protein E16; large neutral amino acids transporter 1; large neutral amino acids transporter small subunit 1; LAT1; LAT-1; L-type amino acid transporter 1; MDU1; MP hLAT1; MPE16; NACAE; Protein TA1; SLC7A5; sodium-independent neutral amino acid transporter LAT1; solute carrier family 7 (amino acid transporter light chain, L system), member 5; solute carrier family 7 (cationic amino acid transporter, y+ system), member 5; solute carrier family 7 member 5; Ta1; tumor-associated protein 1; y+ system cationic amino acid transporter
Rabbit
Protein A
RUO
8140
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.