Learn More
SLC7A5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28670-1-AP-150UL
This item is not returnable.
View return policy
Description
The SLC7A5 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human SLC7A5. This antibody recognizes human antigen. The SLC7A5 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SLC7A5 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| 150 μL | |
| 8140 | |
| IgG |