missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A5/LAT1 Antibody (BU53) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£177.00 - £339.00
Specifications
| Antigen | SLC7A5/LAT1 |
|---|---|
| Clone | BU53 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1 - 2 μg/mL, Immunohistochemistry 1:100 - 1:300, Immunohistochemistry-Paraffin 1:100 - 1:300, Flow (Cell Surface) 1 - 2 μg/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Cell Surface), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18606736
|
Novus Biologicals
NBP2-50465SS |
0.025 mL |
£177.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676108
|
Novus Biologicals
NBP2-50465 |
0.1 mL |
£339.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC7A5/LAT1 Monoclonal antibody specifically detects SLC7A5/LAT1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow (Cell Surface), CyTOFSpecifications
| SLC7A5/LAT1 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Cell Surface), CyTOF | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS | |
| 8140 | |
| IgG2a | |
| Protein G purified |
| BU53 | |
| Flow Cytometry 1 - 2 μg/mL, Immunohistochemistry 1:100 - 1:300, Immunohistochemistry-Paraffin 1:100 - 1:300, Flow (Cell Surface) 1 - 2 μg/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter | |
| Plasma cell line 8226 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title