missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC7A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569257
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Mediates the uptake of the cationic amino acids arginine, lysine and ornithine in a sodium-independent manner.
Specifications
| SLC7A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC7A3 | |
| amino acid transporter, cationic 3; Atrc3; Cat3; CAT-3; Cationic amino acid transporter 3; cationic amino acid transporter y+; SLC7A1; SLC7A2; Slc7a3; solute carrier family 7 (cationic amino acid transporter, y+ system), member 3; solute carrier family 7 member 3; solute carrier family 7, member 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11989 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P70423 | |
| SLC7A3 | |
| synthetic peptide corresponding to a region of mouse SLC7A3. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction