missing translation for 'onlineSavingsMsg'
Learn More

SLC7A10 Antibody, Novus Biologicals™

Product Code. 18347752
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347752 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347752 Supplier Bio-Techne Supplier No. NBP312253

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC7A10 Polyclonal antibody specifically detects SLC7A10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A10
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:4000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:200
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias asc-1, ASC1, asc-type amino acid transporter 1, FLJ20839, HASC-1, solute carrier family 7 (neutral amino acid transporter light chain, asc system), member 10, solute carrier family 7 member 10, solute carrier family 7, (cationic amino acid transporter, y+ system) member 10, solute carrier family 7, (neutral amino acid transporter, y+ system) member 10
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 450-500 on rat Asc-type amino acid transporter 1 protein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56301
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.