missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC6A8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537060
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 70 kDa. Purity is >95% by SDS-PAGE.
SLC6A8 (solute carrier family 6 member 8), also known as CT1 (creatine transporter 1), CRT or CRTR, is a 635 amino acid multi-pass plasma membrane protein that belongs to the sodium/neurotransporter (SNF) family. Expressed in a variety of tissues including kidney, skeletal muscle, heart, brain, prostate, testis and colon, SLC6A8 functions to transport creatine into and out of cells, specifically those found in brain and muscle tissue. Creatine, an organic acid, occurs naturally and is essential for supplying the energy needed for proper muscle and nerve function. Defects in the gene encoding SLC6A8 lead to cerebral creatine deficiency and are the cause of X-linked creatine deficiency syndrome, a rare disorder characterized by facial anomalies, seizures and mental retardation. Multiple isoforms of SLC6A8 exist due to alternative splicing events.
Specifications
| SLC6A8 | |
| Polyclonal | |
| Unconjugated | |
| SLC6A8 | |
| AA589632; CCDS1; choline transporter; CHOT1; CHT1; Creat; creatine transporter; creatine transporter 1; creatine transporter SLC6A8 variant D; creatine transporter, solute carrier family 6, member 8; CRT; CRTR; CT1; CTR5; MGC87396; Na+ dependent transporter; SLC6A8; Sodium- and chloride-dependent creatine transporter 1; solute carrier family 6 (neurotransmitter transporter), member 8; solute carrier family 6 (neurotransmitter transporter, creatine), member 8; solute carrier family 6 member 8; solute carrier family 6, member 8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 102857, 50690, 6535 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P28570, P48029, Q8VBW1 | |
| SLC6A8 | |
| Synthetic peptide corresponding to amino acids 590-635 of human SLC6A8. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction