missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A5, Mouse anti-Human, Clone: 3B3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009152-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: TLFYLFASFVSVLPWGSCNNPWNTPECKDKTKLLLDSCVISDHPKIQIKNSTFCMTAYPNVTMVNFTSQANKTFVSGSEEYFKYFVLKISAGIEYPGEIRSpecifications
| SLC6A5 | |
| Monoclonal | |
| Unconjugated | |
| solute carrier family 6 (neurotransmitter transporter, glycine), member 5 | |
| GLYT2/NET1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3B3 | |
| In 1x PBS, pH 7.4 | |
| NM_004211 | |
| SLC6A5 | |
| SLC6A5 (NP_004202, 294 a.a. ∼ 393 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9152 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction