missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A4/5-HTTLPR/Serotonin transporter Antibody (SR2165), Novus Biologicals™
Rabbit Monoclonal Antibody
£272.00 - £341.00
Specifications
| Antigen | SLC6A4/5-HTTLPR/Serotonin transporter |
|---|---|
| Clone | SR2165 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636314
|
Novus Biologicals
NBP3-21914-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602124
|
Novus Biologicals
NBP3-21914-50ul |
50 μg |
£272.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC6A4/5-HTTLPR/Serotonin transporter Monoclonal antibody specifically detects SLC6A4/5-HTTLPR/Serotonin transporter in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SLC6A4/5-HTTLPR/Serotonin transporter | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 5HT transporter, 5-HTT, hSERT, HTT5-hydroxytryptamine transporter, Na+/Cl- dependent serotonin transporter, OCD1, SERT, sodium-dependent serotonin transporter, solute carrier family 6 (neurotransmitter transporter, serotonin), member 4,5-HTTLPR, Solute carrier family 6 member 4,5HTT | |
| A synthesized peptide derived from human SLC6A4/5-HTTLPR/Serotonin transporter (Uniprot #: P31645) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2165 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6532 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title