missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A4/5-HTTLPR/Serotonin transporter Antibody (632705), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB17292H
This item is not returnable.
View return policy
Description
SLC6A4/5-HTTLPR/Serotonin transporter Monoclonal antibody specifically detects SLC6A4/5-HTTLPR/Serotonin transporter in Human samples. It is validated for Flow Cytometry
Specifications
| SLC6A4/5-HTTLPR/Serotonin transporter | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 632705 | |
| Flow Cytometry | |
| 5HT transporter, 5-HTT, hSERT, HTT5-hydroxytryptamine transporter, Na+/Cl- dependent serotonin transporter, OCD1, SERT, sodium-dependent serotonin transporter, solute carrier family 6 (neurotransmitter transporter, serotonin), member 4,5-HTTLPR, Solute carrier family 6 member 4,5HTT | |
| Human SLC6A4/5-HTTLPR synthetic peptide, Accession # P31645 | |
| 0.1 mL | |
| Neuroscience | |
| 6532 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction