missing translation for 'onlineSavingsMsg'
Learn More

SLC6A3/DAT1 Antibody (mAb16) - Azide and BSA Free, Novus Biologicals™

Product Code. 18327650 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327650 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327650 Supplier Novus Biologicals Supplier No. NBP280954

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC6A3/DAT1 Monoclonal specifically detects SLC6A3/DAT1 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A3/DAT1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone mAb16
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:1000, ELISA, Immunohistochemistry : 1:100, Immunocytochemistry/Immunofluorescence : 1:250, Immunoprecipitation reported in scientific literature (PMID 20643191), Immunohistochemistry-Paraffin 1:100
Formulation Tris-Glycine, 0.15 M NaCl
Gene Alias DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3
Gene Symbols SLC6A3
Host Species Mouse
Immunogen Synthetic peptide corresponding to the N-terminus of rat DAT1. [UniProt# P23977]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6531
Test Specificity Little to no human reactivity has been observed.
Target Species Mouse, Rat, Human (Negative)
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.