missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A19 Rabbit anti-Human, Mouse, Rat, Polyclonal, ProteinTech
Rabbit Polyclonal Antibody
Brand: Proteintech 29372-1-AP-150UL
This item is not returnable.
View return policy
Description
Aliquoting is unnecessary for -20°C storage.
SLC6A19 Polyclonal antibody specifically detects SLC6A19 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SLC6A19 | |
| Polyclonal | |
| Unconjugated | |
| SLC6A19 | |
| B0AT1; HND; SLC6A19; sodium-dependent amino acid transporter system B0; sodium-dependent neutral amino acid transporter B(0)AT1; solute carrier family 6 (neurotransmitter transporter), member 19; solute carrier family 6 (neutral amino acid transporter), member 19; solute carrier family 6 member 19; System B(0) neutral amino acid transporter AT1; system B0 neutral amino acid transporter | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 340024, 664630, 74338 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 700 μg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q2A865, Q695T7, Q9D687 | |
| SLC6A19 | |
| SLC6A19 fusion protein Ag31076 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction