missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC6A18 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA537221

Product Code. 15594301

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody detects endogenous protein at a molecular weight of 71 kDa. Purity is >95% by SDS-PAGE.

The solute carrier (SLC) family, also known as the neurotransmitter transporter family, is one of the largest transporter families in the human genome. The SLC6 subgroup, which includes the dopamine transporter (DAT), the serotonin transporter (SERT) and the norepinephrine transporter (NET), contains sodium/chloride dependent, high-affinity plasma transport proteins. Sodium/chloride-dependent transporter XTRP2, also known as solute carrier family 6 member 18 (SLC6A18), is a 628 amino acid member of the SLC family. XTRP2 is composed of 12 exons, 11 introns and 8 tandem repeats. Highly expressed in the kidney, SLC6A18 is classified as an orphan transporter.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC6A18
Polyclonal
Unconjugated
SLC6A18
b(0)AT3; B0AT3; D630001K16Rik; FLJ31236; Inactive sodium-dependent neutral amino acid transporter B(0)AT3; renal osmotic stress-induced Na-Cl organic solute cotransporter; Rosit; Slc6a18; sodium and chloride dependent transporter Xtrp2; sodium- and chloride-dependent transporter XTRP2; sodium channel-like protein; Sodium-dependent neutral amino acid transporter B(0)AT3; solute carrier family 6 (neurotransmitter transporter), member 18; solute carrier family 6 (neutral amino acid transporter), member 18; solute carrier family 6 member 18; solute carrier family 6, member 18; system B 0 neutral amino acid transporter AT3; system B(0) neutral amino acid transporter AT3; X transporter protein 2; Xt2; Xtrp2
Rabbit
Antigen Affinity Chromatography
RUO
22598, 29323, 348932
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide; pH 7.2
O88576, Q62687, Q96N87
SLC6A18
A synthetic peptide corresponding to residues in human SLC6A18.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.