missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A18 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | SLC6A18 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SLC6A18 Polyclonal antibody specifically detects SLC6A18 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SLC6A18 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 348932 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ31236, Sodium- and chloride-dependent transporter XTRP2, sodium channel-like protein, sodium-dependent neutral amino acid transporter B(0)AT3, solute carrier family 6 (neurotransmitter transporter), member 18, Solute carrier family 6 member 18, solute carrier family 6, member 18, System B(0) neutral amino acid transporter AT3, XTRP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 310-400 of human SLC6A18 (NP_872438.2). VLGFKATNDYEHCLDRNILSLINDFDFPEQSISRDDYPAVLMHLNATWPKRVAQLPLKACLLEDFLDKSASGPGLAFVVFTETDLHMPGAP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title