missing translation for 'onlineSavingsMsg'
Learn More

SLC6A17 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18342053 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342053 100 μg 100µL
18342712 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18342053 Supplier Bio-Techne Supplier No. NBP317542100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC6A17 Polyclonal antibody specifically detects SLC6A17 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A17
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias NTT4, orphan sodium- and chloride-dependent neurotransmitter transporter NTT4, Sodium-dependent neurotransmitter transporter NTT4, solute carrier family 6 (neurotransmitter transporter), member 17, Solute carrier family 6 member 17, solute carrier family 6, member 17
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: RHFHLLSDGSNTLSVSYKKGRMMKDISNLEENDETRFILSKVPSEAPSPMPTHRSYLGPGSTSPLETSGNPNGRYGSGYLLASTPESEL
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388662
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.