missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC6A17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568261
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Functions as a sodium-dependent vesicular transporter selective for proline, glycine, leucine and alanine. In contrast to other members of this neurotransmitter transporter family, does not appear to be chloride-dependent.
Specifications
| SLC6A17 | |
| Polyclonal | |
| Unconjugated | |
| SLC6A17 | |
| D130012J15Rik; MRT48; Ntt4; orphan sodium- and chloride-dependent neurotransmitter transporter NTT4; Rxt1; Slc6a17; Sodium-dependent neurotransmitter transporter NTT4; sodium-dependent neutral amino acid transporter SLC6A17; sodium-dependent orphan neurotramsmitter transporter NTT4; solute carrier family 6 (neurotransmitter transporter), member 17; solute carrier family 6 (neutral amino acid transporter), member 17; solute carrier family 6 member 17; solute carrier family 6, member 17 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 229706, 388662, 613226 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31662, Q8BJI1, Q9H1V8 | |
| SLC6A17 | |
| Synthesized peptide derived from human SLC6A17. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction