missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A16 Monoclonal Antibody (2E5)

SLC6A16 Monoclonal Antibody (2E5)
Brand: Invitrogen MA521753
This item is not returnable.
View return policy
Description
SLC6A16 Monoclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
| SLC6A16 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9GZN6 | |
| SLC6A16 | |
| SLC6A16 (NP_054756.2, 406 a.a. ~ 492 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 2E5 | |
| Unconjugated | |
| SLC6A16 | |
| CTC-301O7.4; FLUP; FREQ; NT5; NTT5; Orphan sodium- and chloride-dependent neurotransmitter transporter NTT5; RGD1561338; Slc6a16; solute carrier family 6 (neurotransmitter transporter), member 16; solute carrier family 6 member 16; solute carrier family 6, member 16 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 28968 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction