missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC6A15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536502
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 85 kDa. Purity is >95% by SDS-PAGE.
SLC6A15 (solute carrier family 6 (neutral amino acid transporter), member 15), also known as sodium-dependent neutral amino acid transporter B(0)AT2, transporter v7-3, NTT73 or sodium-coupled branched-chain amino-acid transporter 1 (SBAT1), is a 730 amino acid multi-pass membrane protein that acts as a sodium-dependent neutral amino acid transporter. A member of the sodium neurotransmitter symporter (SNF) family and SLC6A15 subfamily, SLC6A15 differs from other members of its family in that it does not appear to be chloride-dependent. SLC6A15 is expressed in brain and is encoded by a gene that maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4. 5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.
Specifications
| SLC6A15 | |
| Polyclonal | |
| Unconjugated | |
| Slc6a15 | |
| AA536730; AI326450; AI326451; B0at2; B0AT2 protein; homolog of rat orphan transporter v7-3; hv7-3; NTT73; orphan sodium- and chloride-dependent neurotransmitter transporter NTT73; orphan transporter v7-3; SBAT1; SLC6A15; sodium- and chloride-dependent neurotransmitter transporter NTT73; sodium/chloride dependent neurotransmitter transporter Homo sapiens orphan neurotransmitter transporter NTT7; sodium-coupled branched-chain amino-acid transporter 1; Sodium-dependent neutral amino acid transporter B(0)AT2; solute carrier family 6 (neurotransmitter transporter), member 15; solute carrier family 6 (neutral amino acid transporter), member 15; solute carrier family 6 member 15; solute carrier family 6, member 15; transporter v7-3; V7-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 103098, 282712, 55117 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.2 | |
| Q08469, Q8BG16, Q9H2J7 | |
| Slc6a15 | |
| Synthetic peptide corresponding to amino acids 30-78 of human SLC6A15. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion