missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC6A15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552586
This item is not returnable.
View return policy
Description
Immunogen sequence: RELDDDVTES VKDLLSNEDA ADDAFKTSEL IVDGQEEKDT DVEEGSEVED ERPAWNSKLQ Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.
SLC6A15 (solute carrier family 6 (neutral amino acid transporter), member 15), also known as sodium-dependent neutral amino acid transporter B(0)AT2, transporter v7-3, NTT73 or sodium-coupled branched-chain amino-acid transporter 1 (SBAT1), is a 730 amino acid multi-pass membrane protein that acts as a sodium-dependent neutral amino acid transporter. A member of the sodium neurotransmitter symporter (SNF) family and SLC6A15 subfamily, SLC6A15 differs from other members of its family in that it does not appear to be chloride-dependent. SLC6A15 is expressed in brain and is encoded by a gene that maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4. 5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.
Specifications
| SLC6A15 | |
| Polyclonal | |
| Unconjugated | |
| Slc6a15 | |
| AA536730; AI326450; AI326451; B0at2; B0AT2 protein; homolog of rat orphan transporter v7-3; hv7-3; NTT73; orphan sodium- and chloride-dependent neurotransmitter transporter NTT73; orphan transporter v7-3; SBAT1; SLC6A15; sodium- and chloride-dependent neurotransmitter transporter NTT73; sodium/chloride dependent neurotransmitter transporter Homo sapiens orphan neurotransmitter transporter NTT7; sodium-coupled branched-chain amino-acid transporter 1; Sodium-dependent neutral amino acid transporter B(0)AT2; solute carrier family 6 (neurotransmitter transporter), member 15; solute carrier family 6 (neutral amino acid transporter), member 15; solute carrier family 6 member 15; solute carrier family 6, member 15; transporter v7-3; V7-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55117 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2J7 | |
| Slc6a15 | |
| Recombinant protein corresponding to Human SLC6A15. Recombinant protein control fragment (Product #RP-89570). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction