missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC6A14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587998
This item is not returnable.
View return policy
Description
Immunogen sequence: YYNVIIAYSL YYMFASFQSE LPWKNCSSWS DKNCSRSPIV THCNVSTVNK GIQEIIQMNK SWVDINNFTC INGSEIYQPG QLPSEQYWNK VALQRSSGMN ETGV; Positive Samples: A-549, MCF7, SGC-7901; Cellular Location: Membrane, Multi-pass membrane protein.
This gene encodes a member of the solute carrier family 6. Members of this family are sodium and chloride dependent neurotransmitter transporters. The encoded protein transports both neutral and cationic amino acids.
Specifications
| SLC6A14 | |
| Polyclonal | |
| Unconjugated | |
| SLC6A14 | |
| 1110007A17Rik; 9030613J17Rik; amino acid transporter ATB0+; amino acid transporter B0; amino acid transporter B0+; ATB(0+); Atb0+; ATB0plus; BMIQ11; carrier protein; CATB0plus; colonic system B0+ amino acid transporter; colonic system B0+ amino acid transporter CATB0+; LOW QUALITY PROTEIN: sodium- and chloride-dependent neutral and basic amino acid transporter B(0+); neutral cationic amino acid transporter B0,+; SLC6A14; Sodium- and chloride-dependent neutral and basic amino acid transporter B(0+); solute carrier family 6 (amino acid transporter), member 14; solute carrier family 6 (neurotransmitter transporter), member 14; solute carrier family 6 member 14 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11254, 56774 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.67 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9JMA9, Q9UN76 | |
| SLC6A14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 131-234 of human SLC6A14 (NP_0091621). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction