missing translation for 'onlineSavingsMsg'
Learn More

SLC6A12 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18396792 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396792 100 μL 100µL
18302543 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18396792 Supplier Novus Biologicals Supplier No. NBP305085100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC6A12 Polyclonal specifically detects SLC6A12 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias BGT1, FLJ38727, gamma-aminobutyric acid transporter, solute carrier family 6 (neurotransmitter transporter, betaine/GABA), member 12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 515-614 of human SLC6A12 (NP_003035.3).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6539
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.