missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A12 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | SLC6A12 |
|---|---|
| Clone | 2C1G4 |
| Concentration | 1700 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236420
|
Proteintech
67700-1-IG-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234769
|
Proteintech
67700-1-IG-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SLC6A12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC6A12. This antibody recognizes human, mouse, rat antigen. The SLC6A12 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| SLC6A12 | |
| 1700 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 6539 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2C1G4 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| BGT-1; BGT1; FLJ38727 | |
| IgG2a | |
| Protein A purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title