Learn More
Invitrogen™ SLC6A1 Recombinant Rabbit Monoclonal Antibody (24GB2390)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557260
 View more versions of this product
Product Code. 30276667
  • £126.00 / 20µL
Description
SLC6A1 Recombinant Monoclonal Antibody for Western Blot, Flow
GAT-1 (SLC6A1) The protein encoded by SLC6A1 is a gamma-aminobutyric acid (GABA) transporter that localizes to the plasma membrane. The encoded protein removes GABA from the synaptic cleft, restoring it to presynaptic terminals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC6A1
Recombinant Monoclonal
1.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P31648
SLC6A1
A synthesized peptide derived from human SLC6A1 / GAT1 (aa 1-50).
20 μL
Primary
Mouse
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2390
Unconjugated
SLC6A1
A730043E01; GABA T1; GABA T-1; GABA transporter 1; GABA transporter protein; GABAT1; GABATHG; GABATR; Gabt; GABT1; gamma-aminobutyric acid (GABA-A) transporter 1; GAT1; GAT-1; MAE; SLC6A1; Sodium- and chloride-dependent GABA transporter 1; solute carrier family 6 (neurotransmitter transporter), member 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; solute carrier family 6 member 1; XT-1; Xtrp1
Rabbit
Affinity chromatography
RUO
232333
-20°C