missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC5A8/SMCT1 Antibody (903502), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8398APCCY7
This item is not returnable.
View return policy
Description
SLC5A8/SMCT1 Monoclonal antibody specifically detects SLC5A8/SMCT1 in Human samples. It is validated for Flow Cytometry
Specifications
| SLC5A8/SMCT1 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 160728 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 903502 | |
| Flow Cytometry | |
| AITSolute carrier family 5 member 8, Apical iodide transporter, Electrogenic sodium monocarboxylate cotransporter, MGC125354, SMCTSMCT1, Sodium iodide-related cotransporter, sodium-coupled monocarboxylate transporter 1, solute carrier family 5 (iodide transporter), member 8 | |
| NS0 mouse myeloma cell line transfected with human SLC5A8/SMCT1, Met1-Leu610, Accession # NP_666018 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction