missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC5A8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21433-1-AP-20UL
This item is not returnable.
View return policy
Description
SLC5A8 has been shown to transport iodide by a passive mechanism (Rodriguez et al., 2002 ) and monocarboxylates and short-chain fatty acids by a sodium-coupled mechanism (Gopal et al., 2004 ).Specifications
| SLC5A8 | |
| Polyclonal | |
| Unconjugated | |
| SLC5A8 | |
| AIT, Apical iodide transporter, SLC5A8, SMCT, SMCT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 160728, 216225, 500820 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BYF6, Q8N695 | |
| Slc5a8 | |
| SLC5A8 Fusion Protein Ag16041 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction