missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC5A8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA542514

Product Code. 15505522

  • £440.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Peptide sequence: GILVPFANSI GALVGLMAGF AISLWVGIGA QIYPPLPERT LPLHLDIQGC Sequence homology: Human: 100%; Mouse: 83%; Rabbit: 92%; Rat: 91%.

SLC5A8 has been shown to transport iodide by a passive mechanism (Rodriguez et al., 2002 ) and monocarboxylates and short-chain fatty acids by a sodium-coupled mechanism (Gopal et al., 2004 ).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC5A8
Polyclonal
Unconjugated
Slc5a8
AIT; Apical iodide transporter; apical iodide transporter , putative; electrogenic sodium monocarboxylate cotransporter; RGD1564146; Slc5a8; SMCT; SMCT1; Sodium iodide-related cotransporter; sodium-coupled monocarboxylate transporter 1; solute carrier family 5 (iodide transporter), member 8; solute carrier family 5 (sodium/monocarboxylate cotransporter), member 8; solute carrier family 5 member 8
Rabbit
Affinity Chromatography
RUO
160728
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Surface Plasmon Resonance
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8N695
Slc5a8
Synthetic peptide directed towards the middle region of human SLC5A8.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.