Learn More
Invitrogen™ SLC5A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143934
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, humna RT4 whle cell, rat C6 whole cell, mouse NIH/3T3 whole cell. Flow: JK cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Acetylcholine functions as a neurotransmitter in both the central and peripheral nervous systems of all vertebates, and is the principle neurotransmitter used at the neuromuscular junction. Presynaptic synthesis of acetylcholine requires a steady supply of choline, which is acquired by a plasma membrane high affinity choline transporter (CHT).
Specifications
| SLC5A7 | |
| Polyclonal | |
| Unconjugated | |
| Slc5a7 | |
| CHT; CHT1; hCHT; Hemicholinium-3-sensitive choline transporter; high affinity choline transporter 1; high affinity choline transporter 1; LOW QUALITY PROTEIN: high affinity choline transporter 1; high affinity choline transporter; hemicholinium-3-sensitive choline transporter; HMN7A; MGC126299; MGC126300; similar to solute carrier family 5 (choline transporter), member 7; SLC5A7; solute carrier family 5 (choline transporter), member 7; solute carrier family 5 (sodium/choline cotransporter), member 7; solute carrier family 5 member 7; solute carrier family 5, member 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 60482, 63993, 85426 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q8BGY9, Q9GZV3, Q9JMD7 | |
| Slc5a7 | |
| E.coli-derived human SLC5A7 recombinant protein (Position: R446-Q580). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.