missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC5A6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA557889
This item is not returnable.
View return policy
Description
Immunogen sequence: LNPATIYPVL PKLLSLLPLS CQKRLHCRSY GQDHLDTGLF PEKPRNGVLG DSRDKEAMAL DGTAYQGSSS TCILQET Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 68%.
Transports pantothenate, biotin and lipoate in the presence of sodium. Target protein mediates sodium-dependent transport of pantothenate, biotin, and lipoate; plays a role in intestinal uptake of biotin and in transplacental vitamin transport.
Specifications
| SLC5A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC5A6 | |
| E430023I20; Na(+)-dependent multivitamin transporter; Na+-dependent multivitamin transporter; Slc5a6; SMVT; Sodium-dependent multivitamin transporter; solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6; solute carrier family 5 (sodium-dependent vitamin transporter), member 6; solute carrier family 5 member 6; solute carrier family 5, member 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8884 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y289 | |
| SLC5A6 | |
| Recombinant protein corresponding to Human SLC5A6 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction