missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC5A6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533045
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Rat (100%) Monkey, Mouse, Hamster (95%) Marmoset (89%) Bat, Elephant, Panda, Rabbit, Horse (84%).
Transports pantothenate, biotin and lipoate in the presence of sodium. Target protein mediates sodium-dependent transport of pantothenate, biotin, and lipoate; plays a role in intestinal uptake of biotin and in transplacental vitamin transport.
Specifications
| SLC5A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC5A6 | |
| E430023I20; Na(+)-dependent multivitamin transporter; Na+-dependent multivitamin transporter; Slc5a6; SMVT; Sodium-dependent multivitamin transporter; solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6; solute carrier family 5 (sodium-dependent vitamin transporter), member 6; solute carrier family 5 member 6; solute carrier family 5, member 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 170551, 8884 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O70247, Q9Y289 | |
| SLC5A6 | |
| Synthetic 19 amino acid peptide from N-terminus of human SLC5A6. | |
| 50 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction