missing translation for 'onlineSavingsMsg'
Learn More

SLC5A5/Sodium Iodide Symporter Antibody (902320) [Janelia Fluor™ 585], Novus Biologicals™

Product Code. 30166017 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30166017 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30166017 Supplier Novus Biologicals Supplier No. FAB8367JF585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC5A5/Sodium Iodide Symporter Monoclonal antibody specifically detects SLC5A5/Sodium Iodide Symporter in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A5/Sodium Iodide Symporter
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 902320
Conjugate Janelia Fluor 585
Formulation 50mM Sodium Borate
Gene Alias Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC5A5, Accession # Q92911
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Cellular Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6528
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.