Learn More
SLC5A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21628-1-AP-20UL
 View more versions of this product
Product Code. 16811235
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Prevents intracellular accumulation of high concentrations of myo-inositol (an osmolyte) that result in impairment of cellular function.
Specifications
Specifications
SLC5A3
Polyclonal
Unconjugated
SLC5A3
SLC5A3, SMIT, SMIT2
Rabbit
Antigen Affinity Chromatography
RUO
114507, 53881, 6526
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P53794, Q9JKZ2
Slc5a3
SLC5A3 Fusion Protein Ag16218
20 μL
Primary
Mouse, Human, Rat